ContactUsLinkComponent
VWR® Q Path®, Cytology Funnel
VWR® Q Path®, Cytology Funnel
Catalog # 720-1999
Supplier:  VWR Collection
undefined
VWR® Q Path®, Cytology Funnel
Catalog # 720-1999
Supplier:  VWR Collection
Specifications
  • Description:
    Cytology funnel, Cytocuvette Q Path®, 100 - 800  µl
  • Pk:
    600
Specifications
About this item
Single-use plastic chamber used in cytocentrifugation for urine, gynaecological and non gynaecological specimens, also designed to perform cell suspension transfers on a slide with low risk of contamination.
  • Two apertures with attached filter - large absorption capacity
  • Can resist water and alcohol for 24 H
  • Speed of absorption is ±1 ml in three minutes