VWR® Q Path® Quickspot, Slide Holders for Turbitec®
Supplier: LABONORD
Certificates
About this item
Q Path® Quickspot and slide holder for Turbitec are used to carry out cytocentrifugation. They allow a cell suspension to be deposited on a slide (gynaecological specimens). The slide holder is a support for positioning the slide and Quickspot is the sample filling chamber.
- Q Path® slide holder (reusable) and Q Path® Quickspot (single use)
- Accessories fit together and are used together
- Resistant to centrifugation up to 2039×g for 30 min
Please note: For use with Hettich® centrifuges (Rotofix, Rotina) and compatible rotor and buckets.
Frequently Bought Together