VWR® HistoChoice®, Clearing agent
Supplier: VWR Chemicals
Certificates
About this item
HistoChoice® clearing agent is a much less hazardous replacement for xylene/toluene.
- Faster wax dissolution
- Slower evaporation than xylene with no oily residue
- Compatible with organic mounting media
Designed to work with isopropanol in automated processors.
Frequently Bought Together