ContactUsLinkComponent
Immersion flow cups, 'Ford'
Immersion flow cups, 'Ford'
Catalog # SIMEVF2087
Supplier:  Industrial Physics
undefined
Immersion flow cups, 'Ford'
Catalog # SIMEVF2087
Supplier:  Industrial Physics
Specifications
  • Range:
    70 - 370 cSt
  • Description:
    Viscosity cup, "Ford"
  • Cup orifice:
    4,1 mm
  • Pk:
    1
Specifications
About this item
Titanium anodised cup with a fixed stainless steel nozzle and handle.
Certifications: ASTM D1200
Delivery: Supplied with a hard plastic case.