ContactUsLinkComponent
Dust cloths, standard and high performance
Dust cloths, standard and high performance
Catalog # VILE133909
Supplier:  FHP VILEDA SCS
undefined
Dust cloths, standard and high performance
Catalog # VILE133909
Supplier:  FHP VILEDA SCS
Specifications
  • Packed:
    50/pack
  • Colour:
    Yellow
  • Description:
    High Performance (101)
  • L×W:
    240×600 mm
  • Seq:
    02
  • Pk:
    50
Specifications
About this item
These dust-binding cloths are impregnated with a care oil.
  • Pre-impregnated surface collects dust like a magnet
  • Products retain their effectiveness even after being stored for some time
  • High Performance cloth reduces number of airborne particles
The Standard cloth is ideal for the dry cleaning of hard floor surfaces. The High Performance cloth is ideal for the dry cleaning of hospitals, offices and computer rooms.