Aprons, PVC/polyester
Catalog # DELPTABLIVE
Supplier: DELTA PLUS
Specifications
- Garment application:Industrial
- Gender:Unisex
- Risk area:Rain (waterproof)
- Colour:Green
- L×W:1100×750 mm
- Environmentally Preferable:
- Fabric:PVC coated polyester
- Sleeve or leg length:Short
- Thickness:0,35 mm
- Pk:1
Specifications
About this item
These waterproof aprons are made of PVC coated polyester fabric.
- Drawstring straps
- Double-sided coating
Ideal for non hazardous applications in industries and hygiene environments.
Thickness: 0,35 mm