ContactUsLinkComponent
HPLC columns, Discovery RP-AmideC16, Supelco®
HPLC columns, Discovery RP-AmideC16, Supelco®
Supplier:  Merck
Certificates
HPLC columns, Discovery RP-AmideC16, Supelco®
Supplier:  Merck
Specifications
  • Catalog No:
  • 505013-40
  • 505064-30
  • Carbon load:
  • 11%
  • 11%
  • Endcapped:
  • Yes
  • Yes
  • pH range:
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 200 m²/g
  • 200 m²/g
  • USP code:
  • L60
  • L60