Tubing cutter, Terry-Tool
Cutter for steel tubing.
- Cutting wheel for cutting ¹/₁₆ and ¹/₈" stainless steel tubing
- Clean, right angle cuts with minimal burring or chipping of the tubing
- No additional reaming or deburring is required
: Spare cutting wheels are available.
Frequently Bought Together