Orders LinkContactUsLinkComponent
Microscope slide, UniMark®
Microscope slide, UniMark®
Micoscope slides, UniMark
Catalog # MARI0704022
Supplier:   MARIENFELD
Microscope slide, UniMark®
Catalog # MARI0704022
Supplier:  MARIENFELD
Microscope slide, UniMark®
Microscope slide, UniMark®
Micoscope slides, UniMark
Price
421,91 €
Log in for availability
1 * 2.500 ST
Product Details & Documents
UniMark slide is designed with a white circle of Ø 12 mm and a white marking area of 20 mm for use in cytocentrifuges.
It is suitable for suitable for marking with inkjet of laser printers and permanent markers.
Certifications: DIN 8037/1. IVD 98/79/EC.
Packaging: 50 pieces in a box.
Specifications
  • Description:
    Micoscope slides, UniMark