Antibody saver trays, SP Bel-Art
Antibody saver trays, PP, translucent, minimizes the use of expensive antibodies when incubating filters in the western blot process by using an optimally sized tray.
- Single lane tray in a set of five linked together for easier handling but can be easily separated
- Available for popular midi and mini gels
- Includes attached lid to protect against spillage
- Trays are stackable, enclosable and reusable
Frequently Bought Together