Low pressure tees and crosses, PEEK
Supplier: VICI JOUR RESEARCH
Certificates
About this item
Connectors for low pressure applications are machined from PEEK.
- Flat-bottomed 1/4-28 fitting details
- Completely inert and biocompatible
- Available as the body only, or complete with fittings
Frequently Bought Together