ContactUsLinkComponent
Ultrasonic liquid processors, programmable, Q500 and Q700
undefined
Ultrasonic liquid processors, programmable, Q500 and Q700
  • Catalog No:
  • 432-0237
  • 432-0234
  • 432-0238
  • 432-0233
  • Model:
  • Q700
  • Q500
  • Q700
  • Q500
  • Frequency:
  • 20 kHz
  • 20 kHz
  • 20 kHz
  • 20 kHz
  • Max. output power:
  • 700 W
  • 500 W
  • 700 W
  • 500 W
  • Timer:
  • Up to 72 hours
  • Up to 10 hours
  • Up to 72 hours
  • Up to 10 hours
  • Programmable:
  • 10 memories plus sequencing
  • Time and amplitude
  • 10 memories plus sequencing
  • Time and amplitude
  • W×D×H:
  • 203×387×216 mm
  • 203×387×216 mm
  • 203×387×216 mm
  • 203×387×216 mm
  • Interval time:
  • 1 second to 24 hours
  • 1 second to 1 minute
  • 1 second to 24 hours
  • 1 second to 1 minute
Probes and horns
Frequently Bought Together