Industrial gloves, 377
: Showa
These seamless polyester/nylon knitted gloves feature a nitrile foam coating which protects the hand from oils, hydrocarbons, grease and abrasion. As the gloves are impermeable they are suitable for working in damp or greasy environments.
- Fully coated, extra coating on palm
- Reinforced knit cuff, scalloped edge, anatomical shape
- Robust, great dexterity and good tearing resistance
- Optimal long lasting grip
Ideal for light assembly tasks in construction industries, in the maritime sector and in petrochemical industries.
: EN 388:2016 (4121X); Cat. II
- Catalog No:
- 112-5809
- 112-5812
- 112-5808
- 112-5810
- 112-5811
- Model:
- 377
- 377
- 377
- 377
- 377
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Finish:
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
Frequently Bought Together