Order Entry
Austria
ContactUsLinkComponent
Storage Boxes for Vials, 1,5 ml
Storage Boxes for Vials, 1,5 ml
Catalog # NEOL2-1968
Supplier:  NEOLAB
undefined
Storage Boxes for Vials, 1,5 ml
Catalog # NEOL2-1968
Supplier:  NEOLAB

Specifications

  • Colour:
    Transparent
  • W×D×H:
    275×140×60 mm
  • Pk:
    1

Specifications

About this item

These stackable storage boxes for vials are made of translucent polypropylene (PP).

  • 200 places for 1,5 ml vials
  • Two label holders
  • Box accepts two rack inserts for 1,5 ml microtest tubes