Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes for Vials

Storage Boxes for Vials

Supplier:   NEOLAB
Storage Boxes for Vials
Supplier:  NEOLAB
Certificates
Storage Boxes for Vials
Storage Boxes for Vials

2 Options Available Below

Product Details & Documents

Storage boxes, made of PP, coloured, for HPLC vials, cryotubes or 1,5 and 2,0 ml microcentrifuge tubes with the height of 46 mm and up to 12 mm diameter.

  • Stackable
  • Transparent, tight sealing and hinged lid
  • Also usable as cryobox
  • Autoclavable
Product Family Options

Product Information

  • Compartments
    Colour
    W×D×H ext.
    Availability
    Price
  • 10×10
    Green
    140×153×55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,99 €
    11,99 €
    1 * 1 ST
       11,99 €

  • 10×10
    Blue
    140×153×55 mm
    Discontinued
  • 10×10
    Orange
    140×153×55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,99 €
    11,99 €
    1 * 1 ST
       11,99 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.