Express Eingabe
Austria
ContactUsLinkComponent
Cold trap, Quickfit®
Cold trap, Quickfit®
Catalog # 201-3052
Supplier:  SciLabware
undefined
Cold trap, Quickfit®
Catalog # 201-3052
Supplier:  SciLabware
Specifications
  • Bezeichnung:
    Cold trap head, Pyrex® borosilicate glass, socket size 19/26 NS, cone size 34/35 and 19/26 NS, for vacuum systems
  • Seq:
    01
  • VE:
    1
Specifications
About this item
Designed for the vacuum systems which prevents the vapours from the volatile materials from damaging pumps and easy to use.