ContactUsLinkComponent
Air/steam inlet tubes, plain, Quickfit®
  201-3059
undefined
Air/steam inlet tubes, plain, Quickfit®
  201-3059
  • Ø:
    7 mm
  • Length:
    325 mm
  • Pk:
    5
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Air/steam inlet tubes are designed to fit the range of Quickfit® thermometer/inlet adapters having size 13 thread. Glass tube with jet.