Capillary columns, TraceGOLD™ TG-1MS
These non polar TraceGOLD™ TG-1MS GC columns are ideal for optimal signal-to-noise ratio, sensitivity and MS integrity for a wide variety of applications. These low bleed, stable columns achieve consistent results.
- Non polar, 100% dimethyl polysiloxane
- Equivalent to USP G2
- Exceptionally low bleed
These columns are recommended for hydrocarbons, GROs, drugs of abuse and PCBs.
Frequently Bought Together