ContactUsLinkComponent
Beakers, low form, borosilicate glass, with handle
  SMIT154/600
Beakers, low form, borosilicate glass, with handle
  SMIT154/600
  • Ext.Ø:
    90 mm
  • Capacity:
    600 ml
  • Height:
    125 mm
  • Pk:
    6
Borosilicate glass 3.3, with handle.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for general laboratory use.