Chromatography columns with discs
Supplier: witeg Labortechnik
Certificates
About this item
Chromatography column made of borosilicate glass with fused in sintered glass disc with ST-stopcock at the top with solid plug and screw-thread retaining nut, vacuum connection with drip tip and ST-cone at the bottom.
Frequently Bought Together