Orders LinkContactUsLinkComponent
Assortment Boxes, PP
Assortment Boxes, PP
Supplier:   hünersdorff GmbH
Assortment Boxes, PP
Supplier:  hünersdorff GmbH
Certificates
Assortment Boxes, PP
Assortment Boxes, PP
2 Options Available Below
Product Details & Documents
Assortment box with carrying handle made of tough polypropylene.
  • Coloured bottom made of impact-proof, fracture-proof polypropylene
  • Food-safe colours, use in practically all areas of life possible
  • Resistant to cold and customary acids and alkalis
  • Highly transparent, tightly closing lid avoids mixing of the contents
Product Family Options
Product Information
  • Colour
    W×D×H
    Material
    Availability
    Price
  • 216-3718
    With 8 compartments
    With 8 compartments
    Red
    335×225×55 mm
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,80 €
    15,80 €
    1 * 1 ST
       15,80 €
  • HUEN611900
    With 24 compartments
    With 24 compartments
    Blue
    335×225×55 mm
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,30 €
    18,30 €
    1 * 1 ST
       18,30 €