ContactUsLinkComponent
Microtube racks, reversible, with removable handles
Microtube racks, reversible, with removable handles
Catalog # DELTM-563A
Supplier:  DELTALAB
Microtube racks, reversible, with removable handles
Catalog # DELTM-563A
Supplier:  DELTALAB
Specifications
  • For tube Ø:
    12/8 mm
  • Form:
    Reversible
  • No. of Holes:
    80/60
  • Colour:
    Blue
  • Pk:
    10
Specifications
About this item
PP, reversible.
  • Stackable
  • By removing the handles, units can be firmly anchored laterally to one another
The racks have two sides for different tubes: one side holds up to 80 tubes of 12 mm diameter (for example 10×75 mm or 12×75 mm tubes, microtubes, etc.) and the other side holds up to 60 microtubes, PCR, etc.