Orders LinkContactUsLinkComponent
Digital timer
Digital timer
Digital timer with alarm and instructions in Spanish, English and French, Grey, 64×14×61 mm
Catalog # DELT900600
Supplier:   DELTALAB
Digital timer
Catalog # DELT900600
Supplier:  DELTALAB
Digital timer
Digital timer with alarm and instructions in Spanish, English and French, Grey, 64×14×61 mm
Price
140,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Counts up and down.
  • Capacity: 24 hours
  • Alarm (60 seconds)
  • Incl. clip and small bench stand
Delivery information: Supplied with LR44 battery.
Specifications
  • Colour:
    Grey
  • Description:
    Digital timer with alarm and instructions in Spanish, English and French
  • Seq:
    02
  • W×D×H:
    64×14×61 mm