Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Sep-Pak® Vac
Marketsource
Supplier:   WATERS
Solid phase extraction cartridges, Sep-Pak® Vac
Supplier:  WATERS
Marketsource
Certificates
Solid phase extraction cartridges, Sep-Pak® Vac
6 Options Available Below
Product Details & Documents
These cartridges deliver the option of minimum sample preparation. Consists of moulded, high purity PP syringe-barrel bodies with two PE frits to contain the packing material.
  • Standard male Luer permits use with multi-position vacuum manifolds
  • Colour coded printing on syringe barrels for identification
Product Family Options
Product Information
  • Column Format
    Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • Flash cartridge
    Aminopropyl
    55 - 105 µm
    0,8 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 408,00 €
    408,00 €
    1 * 50 ST
       408,00 €
    Specifications:
    Marketsource
    Marketsource
  • Cartridge
    Aminopropyl
    55 - 105 µm
    0,8 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 348,00 €
    348,00 €
    1 * 50 ST
       348,00 €
    Specifications:
    Marketsource
    Marketsource
  • Cartridge
    C18
    55 - 105 µm
    0,8 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 328,00 €
    328,00 €
    1 * 1 ST
       328,00 €
    Specifications:
    Marketsource
    Marketsource
  • Cartridge
    C18
    55 - 105 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 310,00 €
    310,00 €
    1 * 1 ST
       310,00 €
    Specifications:
    Marketsource
    Marketsource
  • Cartridge
    C18
    55 - 105 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 348,00 €
    348,00 €
    1 * 30 ST
       348,00 €
    Specifications:
    Marketsource
    Marketsource
  • Cartridge
    Silica
    55 - 105 µm
    1,2 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 328,00 €
    328,00 €
    1 * 1 ST
       328,00 €
    Specifications:
    Marketsource
    Marketsource