ContactUsLinkComponent
Disposable particle filtering respirators, FFP2/FFP3, Standard Plus-programm, zetMask®
Disposable particle filtering respirators, FFP2/FFP3, Standard Plus-programm, zetMask®
Catalog # ZVGA30931-94
undefined
Disposable particle filtering respirators, FFP2/FFP3, Standard Plus-programm, zetMask®
Catalog # ZVGA30931-94
Specifications
  • Seq:
    01
  • Pk:
    20
  • With valve:
    +
  • Class:
    FFP2
Specifications
About this item
Ultralight, pre-shaped fine-dust filter masks with exhalation valve for protection against coarse, hazardous particle.
  • High degree of efficiency with minimum space requirements
  • Foam-padded nose area for pressure-free sealed fit
  • Elastic bands
  • Hygienically individually packed; in dispenser box
EN 149:2001
Certifications: EN 149:2001