Orders LinkContactUsLinkComponent
Capillary tubes
Capillary tubes
Supplier:   HILGENBERG
Capillary tubes
Supplier:  HILGENBERG
Certificates
Capillary tubes
9 Options Available Below
Product Details & Documents
Soda-lime glass.
  • Both ends open
Product Family Options
Product Information
  • Length
    Ext.Ø
    Ø int.
    Availability
    Price
  • 30 mm
    0,80 mm
    0,64 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 552,00 €
    552,00 €
    1 * 1.000 ST
       552,00 €
  • 70 mm
    0,55 mm
    0,44 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 531,00 €
    531,00 €
    1 * 1.000 ST
       531,00 €
  • 70 mm
    0,90 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 748,00 €
    748,00 €
    1 * 1.000 ST
       748,00 €
  • 80 mm
    0,50 mm
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 762,00 €
    762,00 €
    1 * 1.000 ST
       762,00 €
  • 80 mm
    0,80 mm
    0,64 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 557,00 €
    557,00 €
    1 * 1.000 ST
       557,00 €
  • 80 mm
    1,00 mm
    0,80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 624,00 €
    624,00 €
    1 * 1.000 ST
       624,00 €
  • 100 mm
    1,00 mm
    0,80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 703,00 €
    703,00 €
    1 * 1.000 ST
       703,00 €
  • 150 mm
    1,00 mm
    0,80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 622,00 €
    622,00 €
    1 * 1.000 ST
       622,00 €
  • 150 mm
    0,90 mm
    0,72 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 590,00 €
    590,00 €
    1 * 1.000 ST
       590,00 €