Glass beads
Supplier: HILGENBERG
Certificates
About this item
Glass beads made of soda lime glass.
- Hydrologic resistance class 3
- Acid resistance class 1
- Alkali resistance class 2
Certifications: Class 3 (according to ISO 719), class 1 (according to DIN 12 116) and class 2 (according to ISO 695).
Frequently Bought Together