Orders LinkContactUsLinkComponent
Pasteur pipettes
Pasteur pipettes
Pasteur pipette, with closed tip and cotton plug, 230 mm, +, 4× 250/box
Catalog # 612-0905
Supplier:   HILGENBERG
Pasteur pipettes
Catalog # 612-0905
Supplier:  HILGENBERG
Pasteur pipettes
Pasteur pipettes
Pasteur pipette, with closed tip and cotton plug, 230 mm, +, 4× 250/box
Price
794,00 €
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents
Pipettes made from soda lime glass.
  • With constriction
  • Available with open tip (unplugged) or with closed tip (with cotton plug)
Spezifikationen
  • Packed:
    4× 250/box
  • Sterile:
    +
  • Description:
    Pasteur pipette, with closed tip and cotton plug
  • Length:
    230 mm
  • Seq:
    01