Order Entry
Austria
ContactUsLinkComponent
NMR tubes, 5 mm, standard
NMR tubes, 5 mm, standard
Catalog # HILG2001745
Supplier:  HILGENBERG
undefined
NMR tubes, 5 mm, standard
Catalog # HILG2001745
Supplier:  HILGENBERG

Specifications

  • Length:
    178 mm
  • Pk:
    50

Specifications

About this item

Borosilicate glass 3,3.

  • One end closed
  • Fire polished edge