Orders LinkContactUsLinkComponent
Waste caps | Avantor
Waste caps
Supplier:   VICI JOUR RESEARCH
Waste caps
Supplier:  VICI JOUR RESEARCH
Certificates
13 Options Available Below
Product Details & Documents
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
Ordering information: Barbed adapters must be ordered separately.
Product Family Options
Product Information
  • Thread
    Connector
    Material
    Availability
    Price
  • JOURJR-S-15303
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    B 53
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 430,00 €
    430,00 €
    1 * 1 ST
       430,00 €
  • JOURJR-S-15313
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    B 53
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 410,00 €
    410,00 €
    1 * 1 ST
       410,00 €
  • JOURJR-S-18304
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    B 83
    4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 575,00 €
    575,00 €
    1 * 1 ST
       575,00 €
  • JOURJR-S-18314
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    B 83
    4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 557,00 €
    557,00 €
    1 * 1 ST
       557,00 €
  • JOURJR-S-11023
    Waste cap, includes O-ring, 1/16" and 1/8" nuts, ferrules and colour-sleeve adapter
    Waste cap, includes O-ring, 1/16" and 1/8" nuts, ferrules and colour-sleeve adapter
    GL 45
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 10M x 1 thread for barbed adapter
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 185,00 €
    185,00 €
    1 * 1 ST
       185,00 €
  • JOURJR-S-18004
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    GLS 80
    4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 430,00 €
    430,00 €
    1 * 1 ST
       430,00 €
  • JOURJR-S-18014
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    GLS 80
    4 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 410,00 €
    410,00 €
    1 * 1 ST
       410,00 €
  • JOURJR-S-16003
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    DIN 60
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 259,00 €
    259,00 €
    1 * 1 ST
       259,00 €
  • JOURJR-S-16013
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    DIN 60
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • JOURJR-S-15103
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    S 51
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 259,00 €
    259,00 €
    1 * 1 ST
       259,00 €
  • JOURJR-S-15113
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    S 51
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • JOURJR-S-16103
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    Waste cap, includes O-ring, 1/4" nuts, ferrules and plug
    S 61
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 1/2-20 thread for 1/4" tubing
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 253,00 €
    253,00 €
    1 * 1 ST
       253,00 €
  • JOURJR-S-16113
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    Waste cap, includes O-ring, nuts, ferrules, plugs and colour-sleeve adapter
    S 61
    3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with M10 x 1 thread for barbed adapter
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
Replacement parts
Product Information
  • Catalog #
    Description
    Material
    Availability
    Price
  • Nut, 1/2"-20, for 1/4" OD tubing
    Delrin®*
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,00 €
    0,00 €
    1 * 1 ST
       0,00 €
Product Information
  • Catalog #
    Material
    Description
    Availability
    Price
  • 20
    Collapsible ferrule, for 1/4" OD tubing
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 0,00 €
    0,00 €
    1 * 1 ST
       0,00 €
Product Information
  • Catalog #
    Description
    Material
    Availability
    Price
  • Barbed adapter, M10 × 1 thread, for 4 mm I.Ø tubing
    PA
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €
Product Information
  • Catalog #
    Description
    Material
    Availability
    Price
  • Barbed adapter, M10 × 1 thread, for 6 mm I.Ø tubing
    PA
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €
Product Information
  • Catalog #
    Description
    Material
    Availability
    Price
  • Plug, 1/4"-28
    PP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,60 €
    3,60 €
    1 * 1 ST
       3,60 €
Product Information
  • Catalog #
    Material
    Description
    Availability
    Price
  • 17,5 mm
    Plug, Ø 10,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,40 €
    13,40 €
    1 * 1 ST
       13,40 €
Product Information
  • Catalog #
    Description
    Material
    Availability
    Price
  • Barbed adapter, M10 × 1 thread, for 8 mm I.Ø tubing
    PA
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.