ContactUsLinkComponent
Cut resistant gloves, Sharpflex Latex
Cut resistant gloves, Sharpflex Latex
Supplier:  HONEYWELL SAFETY
Certificates
undefined
Cut resistant gloves, Sharpflex Latex
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • DALL2232525/10
  • DALL2232525/7
  • DALL2232525/9
  • Model:
  • Latex
  • Latex
  • Latex
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Finish:
  • Crinkled coating on palm and fingertips
  • Crinkled coating on palm and fingertips
  • Crinkled coating on palm and fingertips
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (2X42C), EN 407 (X2XXXX)
  • EN 388:2016 (2X42C), EN 407 (X2XXXX)
  • EN 388:2016 (2X42C), EN 407 (X2XXXX)