ContactUsLinkComponent
Cut resistant gloves, Sharpflex Nit
Cut resistant gloves, Sharpflex Nit
Supplier:  HONEYWELL SAFETY
Certificates
undefined
Cut resistant gloves, Sharpflex Nit
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • DALL2232524/10
  • DALL2232524/11
  • DALL2232524/7
  • DALL2232524/9
  • Model:
  • Nit
  • Nit
  • Nit
  • Nit
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Para-aramid
  • Finish:
  • Foam coating on palm and fingertips
  • Foam coating on palm and fingertips
  • Foam coating on palm and fingertips
  • Foam coating on palm and fingertips
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3X42C), EN 407 (X1XXXX)
  • EN 388:2016 (3X42C), EN 407 (X1XXXX)
  • EN 388:2016 (3X42C), EN 407 (X1XXXX)
  • EN 388:2016 (3X42C), EN 407 (X1XXXX)