ContactUsLinkComponent
Cell detachment solution, Accutase
Catalog # L0950-100
Supplier:  Biowest
CAS Number:  
undefined
Cell detachment solution, Accutase
Catalog # L0950-100
Supplier:  Biowest
CAS Number:  
Specifications
  • Pk:
    100 ml
  • Description:
    Accutase
Specifications
About this item
Accustase is a ready to use cell detachment solution of proteolytic and collagenolytic enzymes. Accutase is a direct gentle replacement for trypsin solution. It can also be used on suspension cells to reduce their agglutination in order to ease counting.