Orders LinkContactUsLinkComponent
Desk timer/stopwatch
Desk timer/stopwatch
Digital timer/stopwatch with jumbo digits, Silver, 160×70×90 mm
Catalog # 609-0002
Marketsource
Supplier:   EA COMBS
Desk timer/stopwatch
Catalog # 609-0002
Supplier:  EA COMBS
Marketsource
Desk timer/stopwatch
Desk timer/stopwatch
Digital timer/stopwatch with jumbo digits, Silver, 160×70×90 mm
Price
181,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Bench top clock/stopwatch with count up/down timer.
  • Four count up timing channels: seconds, minutes, hours; decimal seconds; ¹/₁₀₀ seconds; decimal minutes
  • Countdown timer from 24 hours with memory and alarm at end of period
  • Clock with calendar and daily alarm
  • Accuracy within one second over a 24 hour period
Delivery information: Supplied with R14 battery.
Specifications
  • Colour:
    Silver
  • Description:
    Digital timer/stopwatch with jumbo digits
  • W×D×H:
    160×70×90 mm