Dental clamp forcep, Aesculap®
Catalog # AESCDP152R
Supplier: Aesculap
Specifications
- Length:125 mm
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This angled combination clamp forcep is particularly suitable for positioning brackets in the posterior teeth.
- Mouth parts fluted
- Spindle on forcep end is used for adjusting the brackets