Orders LinkContactUsLinkComponent
Face shield
Face shield
Supplier:   UVEX
Face shield
Supplier:  UVEX
Certificates
Face shield
2 Options Available Below
Product Details & Documents
These polycarbonate and cellulose acetate visors can be attached to helmets by using a length adapter and can be locked in three positions.
  • Robust visor with specially developed adapter for attachment to side helmet slots
  • Extremely lightweight plastic design
  • Material thickness: 1 mm
Ordering information: Visors are supplied with adapter but without helmet. Visors are compatible with helmets uvex airwing.
Product Family Options
Product Information
  • Type
    Material
    Availability
    Price
  • 111-8231
    Face shield, with helmet adapter
    Face shield, with helmet adapter
    Anti-fog
    Cellulose acetate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,10 €
    46,10 €
    1 * 1 ST
       46,10 €
  • UVEX9726.014
    Face shield, with helmet adapter
    Face shield, with helmet adapter
    Uncoated
    Polycarbonate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,50 €
    38,50 €
    1 * 1 ST
       38,50 €
Complementary Products
Product Information
  • Catalog #
    Description
    Colour
    Availability
    Price
  • 111-8232
    Safety helmet, Airwing B
    White
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8235
    Safety helmet, Airwing B
    Yellow
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8216
    Safety helmet, Airwing B
    Orange
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8217
    Safety helmet, Airwing B
    Red
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8218
    Safety helmet, Airwing B
    Green
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8219
    Safety helmet, Airwing B
    Blue
    Log in for availability
    1 * 1 ST - 12,70 €
  • 111-8234
    Safety helmet, Airwing B-WR
    White
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8237
    Safety helmet, Airwing B-WR
    Yellow
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8253
    Safety helmet, Airwing B-WR
    Orange
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8254
    Safety helmet, Airwing B-WR
    Red
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8255
    Safety helmet, Airwing B-WR
    Green
    Log in for availability
    1 * 1 ST - 18,40 €
  • 111-8256
    Safety helmet, Airwing B-WR
    Blue
    Log in for availability
    1 * 1 ST - 18,40 €