Orders LinkContactUsLinkComponent
PCR manual workstation
PCR manual workstation
PCR manual workstation
Supplier:  DELTALAB
Certificates
PCR manual workstation
PCR manual workstation
3 Options Available Below
Product Details & Documents
This storage/working rack, with lid, is designed to work with different PCR volumes at the same time. It can accommodate up to 32 individual 0,2 ml PCR tubes, 24× 1,5/2,0 ml and 16× 0,5 ml microtubes.
  • Tube wells are alphanumerically matrixed with raised bold type for accurate sample identification
  • Rack has a transparent lid for easy viewing of samples
  • Frosted writing panels on four sides allow labelling or writing
  • Supplied with a removable rack accommodating 0,2 ml PCR tubes
Main rack W×D×H: 215×118×50 mm
Removable rack W×D×H: 125×85×15 mm
Documents
Product Family Options
Product Information
  • Availability
    Price
  • 212-9154
    PCR rack with lid, green
    PCR rack with lid, green
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 127,00 €
    127,00 €
    1 * 1 ST
       127,00 €
  • 212-9216
    PCR rack with lid, blue
    PCR rack with lid, blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €
  • 212-9156
    PCR rack with lid, yellow
    PCR rack with lid, yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €