Biopsy processing cassettes, Histosette® II
: DELTALAB
Made of acetal polymer.1 mm square openings to maximise fluid exchange and to ensure proper drainage.
- With 45° angle writing area
- Suitable for automatic labellers
- No need to break the lid and then put it in place, as it is supplied pre-mounted
- Can be easily opened and closed with just one hand
Frequently Bought Together