Orders LinkContactUsLinkComponent
Test tubes, round bottom
Test tubes, round bottom
Test tubes, round bottom
Supplier:  DELTALAB
Certificates
Test tubes, round bottom
9 Options Available Below
Product Details & Documents
Tubes are made of borosilicate or soda glass.
  • Good chemical resistance
  • Good temperature resistance
High quality of tubes is reflected in uniformity of wall thickness, diameter and height dimensions.
Documents
Product Family Options
Soda glass
Product Information
  • Capacity
    Length
    Ext.Ø
    Wall thickness
    Ø int.
    Availability
    Price
  • 4 ml
    75 mm
    9,8 mm
    0,60 mm
    8,2 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 151,00 €
    151,00 €
    1 * 1.000 ST
       151,00 €
  • 6 ml
    75 mm
    11,6 mm
    0,60 mm
    10,2 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 48,80 €
    48,80 €
    1 * 250 ST
       48,80 €
  • 22 ml
    150 mm
    16 mm
    0,70 mm
    13,6 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 102,00 €
    102,00 €
    1 * 250 ST
       102,00 €
  • 28 ml
    150 mm
    18 mm
    0,85 mm
    15 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 135,00 €
    135,00 €
    1 * 250 ST
       135,00 €
  • 34 ml
    150 mm
    20 mm
    0,85 mm
    17,2 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 105,00 €
    105,00 €
    1 * 100 ST
       105,00 €
Borosilicate glass
Product Information
  • Capacity
    Length
    Ext.Ø
    Wall thickness
    Ø int.
    Availability
    Price
  • 6 ml
    75 mm
    11,6 mm
    0,60 mm
    10,2 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 61,40 €
    61,40 €
    1 * 250 ST
       61,40 €
  • 10 ml
    100 mm
    12,7 mm
    0,60 mm
    11,1 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 79,50 €
    79,50 €
    1 * 250 ST
       79,50 €
  • 15 ml
    100 mm
    15,8 mm
    0,60 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 473,00 €
    473,00 €
    1 * 1.000 ST
       473,00 €
  • 22 ml
    150 mm
    16 mm
    0,70 mm
    13,6 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 123,00 €
    123,00 €
    1 * 250 ST
       123,00 €