Orders LinkContactUsLinkComponent
Sample container with screw cap | Avantor
Sample container with screw cap
Sample container with screw cap
Supplier:  DELTALAB
Certificates
Sample container with screw cap
Sample container with screw cap
6 Options Available Below
Product Details & Documents
Container PP, natural or blue, with PE screw cap.
  • With or without label
  • Blue containers are ideal for photosensitive samples
  • Bulk or individually wrapped
To store and transport samples securely.
Achtung! Bitte beachten: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Type
    Colour
    Cap colour
    Packed
    Availability
    Price
  • 60 ml
    38 mm
    65 mm
    With white label
    Blue
    Blue
    Bulk
    Log in for availability
    Log in for availability
    1 * 600 ST
    1 * 600 ST - 255,00 €
    255,00 €
    1 * 600 ST
       255,00 €
  • 150 ml
    57 mm
    73 mm
    With label
    Clear
    Red
    Bulk
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 241,00 €
    241,00 €
    1 * 450 ST
       241,00 €
  • 150 ml
    57 mm
    73 mm
    Sterile R (irradiated)
    Clear
    Red
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 350 ST
    1 * 350 ST - 265,00 €
    265,00 €
    1 * 350 ST
       265,00 €
  • 150 ml
    57 mm
    73 mm
    Sterile A (aseptic)
    Clear
    Red
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 350 ST
    1 * 350 ST - 122,00 €
    122,00 €
    1 * 350 ST
       122,00 €
  • 150 ml
    57 mm
    73 mm
    Without label
    Clear
    Red
    Bulk
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 212,00 €
    212,00 €
    1 * 450 ST
       212,00 €
  • 216-4052
    Ideal for photosensitive samples
    Ideal for photosensitive samples
    150 ml
    57 mm
    73 mm
    With white label
    Blue
    Blue
    Bulk
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 260,00 €
    260,00 €
    1 * 450 ST
       260,00 €