Order Entry
Austria
ContactUsLinkComponent
 
General laboratory tool kit
Catalog # HEWL5180-4162
Supplier:  Agilent
undefined
General laboratory tool kit
Catalog # HEWL5180-4162
Supplier:  Agilent

Specifications

  • Description:
    General laboratory tool kit
  • Pk:
    1

Specifications

About this item

The general laboratory tool kit is made in the USA and includes the following items:

  • Toolbox
  • Flashlight
  • 8" bastard file
  • 6" adjustable wrench
  • 12" adjustable wrench for use with regulator
  • No.1 pt, 3" Pozidriv shaft, fits no. 2 to 4 screws
  • No.2 pt, 4" Pozidriv shaft, fits no. 5 to 10 screws
  • Flathead screwdriver, 2" steel shaft, ¹/₈" blade
  • Flathead screwdriver, 4" steel shaft, ¹/₄" blade
  • Hex key set, 1,5; 2; 2,5; 3; 4; 5; and 6 mm keys
  • 6" long jaw needle nose pliers with side cutters
  • Three open-end wrenches (¹/₄×⁵/₁₆", ¹/₂×⁹/₁₆", ⁷/₁₆׳/₈")
  • Snoop-liquid leak detector
  • PTFE tape