Orders LinkContactUsLinkComponent
Test tubes,with screw thread
Test tubes,with screw thread
Test tubes,with screw thread
Supplier:  DELTALAB
Certificates
Test tubes,with screw thread
6 Options Available Below
Product Details & Documents
These screw threaded glass tubes are made of borosilicate or soda glass.
  • Polypropylene caps are designed for the round bottom tubes
  • Aluminium caps are designed for flat bottom tubes
  • Resistant to 4000 min⁻¹
Delivery information: Supplied in small quantities per case for a more convenient use in laboratory. Tubes are supplied without caps.
Documents
Product Family Options
Round bottom
Product Information
  • Capacity
    Length
    Ext.Ø
    Thread
    Wall thickness
    Ø int.
    Availability
    Price
  • 5 ml
    100 mm
    12,8 mm
    13-415
    1,0 mm
    10,7 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 998,00 €
    998,00 €
    1 * 1.000 ST
       998,00 €
  • 11 ml
    100 mm
    16 mm
    15-415
    1,05 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 1 340,00 €
    1 340,00 €
    1 * 1.000 ST
       1 340,00 €
  • 14 ml
    125 mm
    16 mm
    15-415
    1,05 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 403,00 €
    403,00 €
    1 * 250 ST
       403,00 €
  • 18 ml
    150 mm
    16 mm
    15-415
    1,05 mm
    14 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 502,00 €
    502,00 €
    1 * 250 ST
       502,00 €
  • 34 ml
    125 mm
    20 mm
    18-415
    1,15 mm
    17,7 mm
    Log in for availability
    Log in for availability
    1 * 125 ST
    1 * 125 ST
Flat bottom
Product Information
  • Capacity
    Length
    Ext.Ø
    Thread
    Wall thickness
    Ø int.
    Availability
    Price
  • 24 ml
    17,8 mm
    15-415
    1,05 mm
    14,2 mm
    Log in for availability
    Log in for availability
    1 * 252 ST
    1 * 252 ST - 288,00 €
    288,00 €
    1 * 252 ST
       288,00 €
Accessories
Product Information
  • Catalog #
    Material
    Thread
    Availability
    Price
  • DELT617000
    Aluminium
    15-415
    Log in for availability
    1 * 500 ST - 342,00 €
Product Information
  • Catalog #
    Material
    Thread
    Availability
    Price
  • DELT617300
    PP
    18-415
    Log in for availability
    1 * 500 ST - 284,00 €
Product Information
  • Catalog #
    Material
    Thread
    Availability
    Price
  • DELT617200
    PP
    15-415
    Log in for availability
    1 * 1.000 ST - 355,00 €
Product Information
  • Catalog #
    Material
    Thread
    Availability
    Price
  • DELT617100
    PP
    13-415
    Log in for availability
    1 * 1.000 ST - 361,00 €