Sampling spoons, eco-friendly, SP Bel-Art
PLA (polylactic acid) resin, eco-friendly, white.
- Green alternative to petroleum-based sampling tools; helps meet sustainability goals
- FDA grade spoons are safe for food, drug and cosmetic contact
- Comparable in strength and durability to PS, PP and other plastic resin sampling tools
- Long handle (17,8 cm) helps prevent personal contact with sampled material when reaching into bottles, jars and other confined areas
- Rim of each bowl has a flat edge to assist in collecting and measuring small amounts with accuracy
The production of 100% plant-based PLA contributes less greenhouse gasses and uses less non renewable energy from source to resin than its petroleum-based counterparts. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill. Use a separate spoon for each material sampled to avoid cross contamination.
- Catalog No:
- BELAF369401000
- BELAF369421000
- BELAF369441000
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
Frequently Bought Together