Orders LinkContactUsLinkComponent
Digital block reactor, DRB 200
Digital block reactor, DRB 200
 :   Hach
Digital block reactor, DRB 200
 :  Hach
Digital block reactor, DRB 200
Digital block reactor, DRB 200
Digital block heaters which can hold one or two heating blocks, units allow cuvettes and reaction vessels to be digested at identical or different temperatures and time settings. Heating blocks enable cuvettes and reaction vessels to be digested at identical or different temperatures and time settings.
  • Pre-programmed for all standard digestions and freely programmable for user-specific digestions
  • Temperature stability
  • Excellent reproducibility
  • Simple handling, easy and safe to operate
  • Large, easily readable display and one-key operation for standard digestions
Documents
Product Family Options
  • Type
    No. of Holes
    No. of blocks
    Temperature Range
    Temp. stability
    W×D×H
    Weight
  • DRB 200-1
    9×16 mm Ø
    2×20 mm Ø
    1
    +37...+150 °C
    ±1 °C
    250×310×145 mm
    2 kg
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €
  • DRB 200-1
    15×16 mm Ø
    +37...+150 °C
    ±1 °C
    250×310×145 mm
    2 kg
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €
  • DRB 200-2
    21×16 mm Ø
    4×20 mm Ø
    2
    +37...+150 °C
    ±1 °C
    250×310×145 mm
    2,8 kg
    1 * 1 ST
    1 * 1 ST - 2 700,00 €
    2 700,00 €
    1 * 1 ST
       2 700,00 €
  • DRB 200-2
    30×16 mm Ø
    (15 on each of the 2 blocks)
    2
    +37...+150 °C
    ±1 °C
    250×310×145 mm
    2,8 kg
    1 * 1 ST
    1 * 1 ST - 2 700,00 €
    2 700,00 €
    1 * 1 ST
       2 700,00 €
Accessories
  •  
    Description
  • HACHHHA155
    Reducing adapter for DRB200, converts 20 mm diameter vial wells to 16 mm diameter
    1 * 1 ST - 16,00 €