Orders LinkContactUsLinkComponent
Spine model
Spine model
Spine model, 1,8 kg
Catalog # BINH1000125
Marketsource
Supplier:   3B Scientific
Spine model
Catalog # BINH1000125
Supplier:  3B Scientific
Marketsource
Spine model
Spine model
Spine model, 1,8 kg
Price
424,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
In addition to the classic spine, this spine has a sacral opening and exposed brain stem for advanced studies. Other features are:
  • Full pelvis and occipital plate
  • L3-L4 disc prolapsed on spinal column
  • Spinal nerve exits
  • Cervical vertebral artery
  • Male pelvis
Suitable for studying the anatomy of the human spine.
Ordering information: Stand is not included.
Specifications
  • Weight:
    1,8 kg
  • Description:
    Spine model