ContactUsLinkComponent
Insulated Box with Lid
Insulated Box with Lid
Catalog # STOR505710
Supplier:  STOROPACK
Insulated Box with Lid
Catalog # STOR505710
Supplier:  STOROPACK
Specifications
  • W×D×H ext.:
    350×350×300 mm
  • Capacity:
    12,5 l
  • Pk:
    4
Specifications
About this item
Insulated EPS boxes with lid, suitable for the transport of packaged food and ideal for pharmaceutical products.