Order Entry
Austria
ContactUsLinkComponent
Pipette controller, Pipette Helper®
Pipette controller, Pipette Helper®
Catalog # 613-2940
Supplier:  EPPENDORF
   Low Manufacturing Impact
undefined
Pipette controller, Pipette Helper®
Catalog # 613-2940
Supplier:  EPPENDORF
   Low Manufacturing Impact

Specifications

  • Description:
    Pipette Helper®
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Pk:
    1

Specifications

About this item

Pipette controller for glass and plastic pipettes from 0,1 to 100 ml.

  • Aspiration bulb provides rapid filling of the pipette (up to 100 ml at once)
  • Ergonomic lever provides gentle control of aspiration and dispensing
  • Drop wise dispensing and blow out without splashing
  • Drip stop via hydrophobic membrane filter (3 µm)
  • Easy maintenance and cleaning, fully autoclavable

The lightweight Pipette Helper® is the perfect instrument for inexperienced users as its robust and intuitive design allows anyone to adjust the meniscus precisely. The valve unit is optimised so that liquids can be drawn up simply, without exerting pressure. The specially designed aspirating cone ensures secure seating for all measuring and volumetric pipettes (glass and plastic).