Baumé hydrometers
Supplier: Amarell
Jump to:
About this item
General purpose Baumé hydrometers with or without thermometer.
- Reference temperature of 15 °C
- Paper scale with visible print
- Shot weighted body
Colourless round capillary with red or blue filling.
19 Options Available Below
Product Details & Documents
General purpose Baumé hydrometers with or without thermometer.
- Reference temperature of 15 °C
- Paper scale with visible print
- Shot weighted body
Colourless round capillary with red or blue filling.
Product Family Options
Product Information
- RangeDivisionLengthAvailabilityPrice