Orders LinkContactUsLinkComponent
Temperature probe
Immersion probe, PT 100, class B, non-corrosive stainless steel with plastic handle and one-meter four-wire PVC cable, −50…+400 °C, 3×150 mm
  620-4728
 :   Greisinger
Temperature probe
  620-4728
Temperature probe
Temperature probe
Immersion probe, PT 100, class B, non-corrosive stainless steel with plastic handle and one-meter four-wire PVC cable, −50…+400 °C, 3×150 mm
1 * 1 ST
  • Range:
    −50…+400 °C
  • Description:
    Immersion probe, PT 100, class B, non-corrosive stainless steel with plastic handle and one-meter four-wire PVC cable
  • Ø×L:
    3×150 mm