ContactUsLinkComponent
Deep well microplates, 96-round well
Deep well microplates, 96-round well
Catalog # 732-2479
Supplier:  BIOTIX
undefined
Deep well microplates, 96-round well
Catalog # 732-2479
Supplier:  BIOTIX
Specifications
  • Sterile:
    +
  • Colour:
    Clear
  • Description:
    Deep well microplate, 96-round well with U-bottom
  • Seq:
    01
  • Pk:
    5
  • Well volume:
    1,2 ml
Specifications
About this item
Medical grade, virgin PP, untreated. With a smooth, round-well geometry, these deep well microplates maximise sample volume while eliminating the 'wicking' effect common with square-well microplates.
  • Excellent chemical and temperature resistance
  • Tested free from DNase, RNase and endotoxin
Certifications: Conform to ANSI/SBS 1-2004 standards for automated workstations. All plates are BioReady™ certified for purity.